Skip to Content
Merck

Skip To

C-14029

Human Cardiac Microvascular Endothelial Cells (HCMEC)

1 million cells per pellet

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
9 320,00 CZK

About This Item

NACRES:
NA.81
UNSPSC Code:
41106514
Biological source:
human heart (ventricles)
Morphology:
(endothelial)
Relevant disease(s):
diabetes; hypertension; ischemia/reperfusion injury

9 320,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human heart (ventricles)

packaging

pkg of 1,000,000 cells

morphology

(endothelial)

application(s)

cell analysis (DNA, RNA, and protein)

relevant disease(s)

diabetes; hypertension; ischemia/reperfusion injury

shipped in

dry ice

storage temp.

−20°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details. Cells are dissolved in RNAlater for subsequent RNA, DNA, or protein analysis. Note that cell pellets cannot be revived.

Application

Primary Human Cardiac Microvascular Endothelial Cells (HCMEC) are isolated from heart ventricles from a single donor. The cells are routinely analyzed by immunofluorescent staining: they stain positive for CD31 and Willebrand factor and negative for smooth muscle alpha-actin.HCMEC intensively interact with cardiomyocytes and therefore have a characteristic phenotype different from other microvascular endothelial cells. HCMEC play an important role in the physiological regulation of coronary blood flow and capillary exchange. Medical conditions such as hypertrophy, ischemia, hypertension, and diabetes mellitus are associated with endothelial dysfunction.

Biochem/physiol Actions

Heart

Physical form

Pellet is dissolved in RNAlater® for downstream analysis of DNA, RNA or protein profiles. Cells are not revivable.

Preparation Note

Click here for more information.

Analysis Note

Rigid quality control tests are performed for each lot of the corresponding cells. They are tested for cell viability, cell morphology, growth performance and for cell-type specific markers. In addition, all cells have been tested for the absence of HIV-1, HIV-2, HBV, HCV, HTLV-1, HTLV-2 and microbial contaminants (fungi, bacteria, and mycoplasma).

Other Notes

The Cell Pellets can be stored indefinitely at -20°C, and for up to 1 month at 4°C. Note: The Cell Pellets will not freeze at -20°C. Always centrifuge product briefly before opening vial.

Legal Information

RNAlater is a registered trademark of Thermo Fisher Scientific, Inc.

Disclaimer

Although tested negative for HIV-1, HIV-2, HBV, HCV, HTLV-1 and HTLV-2, the cells – like all products of human origin – should be handled as potentially infectious. No test procedure can completely guarantee the absence of infectious agents.
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C12285C12286C12222
description

1 million cells per pellet

description

500,000 cryopreserved cells

description

500,000 proliferating cells

description

500,000 proliferating cells

morphology

(endothelial)

morphology

( endothelial)

morphology

( endothelial)

morphology

( endothelial)

relevant disease(s)

diabetes; hypertension; ischemia/reperfusion injury

relevant disease(s)

diabetes; hypertension; ischemia/reperfusion injury

relevant disease(s)

diabetes; hypertension; ischemia/reperfusion injury

relevant disease(s)

-

biological source

human heart (ventricles)

biological source

human heart (ventricles)

biological source

human heart (ventricles)

biological source

human coronary artery

shipped in

dry ice

shipped in

dry ice

shipped in

ambient

shipped in

ambient

storage temp.

−20°C

storage temp.

−196°C

storage temp.

room temp

storage temp.

room temp

packaging

pkg of 1,000,000 cells

packaging

pkg of 500,000 cells

packaging

pkg of 500,000 cells

packaging

pkg of 500,000 cells


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

7783-20-2

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters