Skip to Content
Merck

Skip To

A4611

AMPK (α1/β2/γ1), active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

AMPKG, MGC33776, MGC57364, MGC61468, MGC8666, PRKAA1, PRKAB2, PRKAG1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
10 μg
Please contact Customer Service for Availability
CZK 27,900.00

About This Item

NACRES:
NA.32
UNSPSC Code:
51111800
Specific activity:
297-401 nmol/min·mg
Assay:
≥70% (SDS-PAGE)
Recombinant:
expressed in baculovirus infected Sf9 cells

CZK 27,900.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in baculovirus infected Sf9 cells

Quality Segment

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

297-401 nmol/min·mg

mol wt

A1 subunit ~68 kDa, B2 subunit ~36 kDa, G1 subunit ~40 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Biochem/physiol Actions

AMPK (A1/B2/G1) is a member of the AMPK family which are heterotrimeric proteins consisting of an α catalytic subunit, and non-catalytic β and γ subunits. AMPKs are an important energy-sensing enzyme group in the cells that monitor energy status particularly in response to stress. AMPKs regulate fatty acid and cholesterol synthesis by regulating the key rate-limiting enzymes acetyl-CoA carboxylase and 3-hydroxy-3-methylglutaryl-CoA reductase. The β subunit may be a positive regulator of AMPK activity and is highly expressed in skeletal muscle.

Physical form

Supplied in 50 mM Sodium Phosphate, pH 7.0, with 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A1233A4486SRP5004
Gene Information

human ... PRKAA1(5562), PRKAB2(5565), PRKAG1(5571)

Gene Information

human ... PRKAA1(5562), PRKAB1(5564), PRKAG1(5571)

Gene Information

human ... PRKAA1(5562), PRKAB1(5564), PRKAG3(53632)

Gene Information

human ... PRKAA2(5563), PRKAB2(5565), PRKAG2(51422)

assay

≥70% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

assay

≥70% (SDS-PAGE)

specific activity

297-401 nmol/min·mg

specific activity

626-848 nmol/min·mg

specific activity

157-213 nmol/min·mg

specific activity

255-345 nmol/min·mg

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in baculovirus infected Sf9 cells

recombinant

expressed in baculovirus infected Sf9 cells

mol wt

A1 subunit ~68 kDa, G1 subunit ~40 kDa, B2 subunit ~36 kDa

mol wt

A1 subunit ~68 kDa, B1 subunit ~38 kDa, G1 subunit ~40 kDa

mol wt

A1 subunit ~68 kDa, B1 subunit ~38 kDa, G3 subunit ~51 kDa

mol wt

~36 kDa (B2), ~65 kDa (G2), ~69 kDa (A2)


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
A4611-10UG04061832847474

Questions

Reviews

No rating value

Active Filters