29625-U
SGE autosampler syringes, fixed needle, for CTC/LEAP
10F-C/F-5/0.47C, Slim line, fixed needle, volume 10 μL, needle size 26 ga, needle L × O.D. 50 mm × 0.47 mm
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
Doporučené produkty
material
cone tip stainless steel needle
needle size
26 ga
description
color (dark orange)
feature
point style 1
syringe type F (cemented needle)
packaging
pkg of 1 ea
manufacturer/tradename
SGE 002980
needle L × O.D.
50 mm × 0.47 mm
volume
10 μL
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)