Key Documents
21033
Supelco
Filter Cartridge
About This Item
UNSPSC Code:
12190000
packaging
pkg of 1 ea
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
General description
Filter Cartridge consists of 1 O-ring, 1 filter cartridge body, 1 filter ring, and 1 cartridge screw cap.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)