Změnit zobrazení
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
26 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 28 100,00 Kč |
O této položce
NACRES:
NB.22
UNSPSC Code:
41122105
Capacity:
600 mL
Packaging:
pack of 26 ea
28 100,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlipackaging
pack of 26 ea
manufacturer/tradename
TPP 87600
capacity
600 mL
size
General description
TPP® TubeSpin bioreactor bottles are designed for large-scale screening, optimization, and process of cell suspension system. These bioreactor bottles offer several advantages, including the ability to evaluate small amounts of important parameters during production upscaling, thereby maximizing the number of parallel experiments at the same time. This approach effectively lowers cost and increases the efficiency of the process.
Features and Benefits
- Conical bottom with filter cap
- Fit standard centrifuge apparatus
Legal Information
TPP is a registered trademark of TPP Techno Plastic Products AG
1 of 1
Tato položka | |||
|---|---|---|---|
| description capacity 600 mL | description filter capacity 250 mL, pack of 12 ea | description capacity 50 mL, polypropylene, conical bottom tube | description capacity 50 mL, polypropylene |
| manufacturer/tradename TPP 87600 | manufacturer/tradename TPP 99250 | manufacturer/tradename TPP 91050 | manufacturer/tradename TPP 91051 |
| capacity 600 mL | capacity - | capacity 50 mL | capacity 50 mL |
| packaging pack of 26 ea | packaging pack of 12 ea | packaging pack of 360 ea | packaging pack of 320 ea |
| size
| size 93 mm × 93 mm × 143 mm | size - | size - |
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?