Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
320 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 9 790,00 Kč |
O této položce
9 790,00 Kč
material
cap, polypropylene , transparent tube, conical bottom tube (with rim)
sterility
sterile
packaging
pack of 320 ea
manufacturer/tradename
TPP 91051
capacity
50 mL
General description
Temperature Range for Polypropylene Tubes: -196°C to +121°C; Polystyrene Tubes: -40°C to +70°C; Maximum RCF for Polypropylene Tubes: 15500 × g*; Polystyrene Tubes: 1700 × g*.
* Depending on adapter and temperature
All Tubes are gamma Irradiated
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description capacity 50 mL, polypropylene | description capacity 15 mL, polypropylene | description capacity 15 mL, polystyrene, conical | description capacity 50 mL, polypropylene, conical bottom tube |
| manufacturer/tradename TPP 91051 | manufacturer/tradename TPP 91015 | manufacturer/tradename TPP 91115 | manufacturer/tradename TPP 91050 |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| material cap, transparent tube, polypropylene , conical bottom tube (with rim) | material polypropylene , conical bottom tube, transparent tube, cap | material transparent tube, polystyrene , conical bottom tube, cap | material polypropylene , cap, conical bottom tube, transparent tube |
| capacity 50 mL | capacity 15 mL | capacity 15 mL | capacity 50 mL |
| packaging pack of 320 ea | packaging pack of 800 ea | packaging pack of 800 ea | packaging pack of 360 ea |
Still not finding the right product?
Explore all of our products under TPP® centrifuge tubes
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?