Změnit zobrazení
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
180 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 14 000,00 Kč |
O této položce
NACRES:
NB.22
UNSPSC Code:
41121706
14 000,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlipackaging
pack of 180 ea
manufacturer/tradename
TPP 87050
capacity
50 mL
size
General description
TPP® TubeSpin bioreactor tubes are ideal for large scale screening and optimizing of suspension cell system process development. The crucial parameters for the production upscaling can be analyzed in small amounts. This makes it cost-effective and increases the process efficiency.
Features and Benefits
- Available in conical or round bottom.
- Filter cap with partly 5 different openings (A, B, C, D, E) or 10 equal openings.
- Centrifuge apparatus which is fit standard.
- Spin: 15′500 x g or 3′500 x g.
- Provides matching adapter plates for shakers for the cultivation of suspension cells.
Legal Information
T3P is a registered trademark of Archimica GmbH
TPP is a registered trademark of TPP Techno Plastic Products AG
1 of 1
Tato položka | |||
|---|---|---|---|
| description capacity 50 mL | description capacity 50 mL, polypropylene | description capacity 15 mL, polystyrene, conical | description capacity 15 mL, polypropylene |
| size
| size - | size - | size - |
| capacity 50 mL | capacity 50 mL | capacity 15 mL | capacity 15 mL |
| manufacturer/tradename TPP 87050 | manufacturer/tradename TPP 91051 | manufacturer/tradename TPP 91115 | manufacturer/tradename TPP 91015 |
| packaging pack of 180 ea | packaging pack of 320 ea | packaging pack of 800 ea | packaging pack of 800 ea |
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?