Key Documents
Z720178
Hettich® centrifuge rotors
for ROTINA 38/38R Centrifuge, Fixed Angle Rotor, 6 places
About This Item
UNSPSC Code:
41103900
NACRES:
NB.22
feature
RCF 13,640 ‑ 16,504
capacity 1.5 ‑ 94 mL
lid
manufacturer/tradename
Hettich 1792
technique(s)
centrifugation: suitable
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG