Hlavní dokumenty
Z717398
Nichiryo pipette tips
maximum volume 5 mL, universal, bulk pack
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
colorless
sterility
non-sterile
feature
DNase free
RNase free
barrier: no
packaging
pack of 100 ea (Bag)
manufacturer/tradename
Nichiryo BMT-XR
availability
not available in (France)
maximum volume
5 mL
Features and Benefits
  • RNase and DNase free
  • Fits most pipettes
  • Premium packaging featuring stand-up and puncture-resistant bags
  • Racked tips are offered in both hinged and lift-off tops
Other Notes
Browse all pipette tips