Z652164
Hettich® MIKRO 200/200R centrifuge accessories
for MIKRO 200/200R Centrifuge, Fixed Angle Rotor, 4 place, for PCR strips
About This Item
Doporučené produkty
feature
RCF 12,490
capacity 12,490 mL
manufacturer/tradename
Hettich 2418
technique(s)
PCR: suitable
centrifugation: suitable
General description
E2040 (Z652172) is an optional lid for rotor 2418 (Z652164)
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Required but not provided
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)