Hlavní dokumenty
Z645532
Nichiryo pipette tips
maximum volume 200 μL, universal, pack of 960 (10 racks of 96 tips), sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
sterility:
sterile
feature:
barrier: no
packaging:
pack of 960 (10 racks of 96 tips)
manufacturer/tradename:
Nichiryo BMT-SGRS
material
colorless
sterility
sterile
feature
barrier: no
packaging
pack of 960 (10 racks of 96 tips)
manufacturer/tradename
Nichiryo BMT-SGRS
availability
not available in (France)
maximum volume
200 μL
Features and Benefits
  • RNase and DNase free
  • Fits most pipettes
  • Premium packaging featuring stand-up and puncture-resistant bags
  • Racked tips are offered in both hinged and lift-off tops
Other Notes
Browse all pipette tips
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)