Key Documents
Z380202
Test tube half racks for orbital mixer
16-mm tubes 5 × 6 array
About This Item
UNSPSC Code:
41122804
NACRES:
NB.73
feature
Holds 25 x 16 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18748-0016
size
128 mm × 105 mm × 72 mm
Application
For secure orbital mixing of samples in test tubes.