Hlavní dokumenty
Z337285
BRAND® conical joint stopper, PP
joint: ST/NS 24/29
About This Item
UNSPSC Code:
41122500
NACRES:
NB.35
joint
joint: ST/NS24/29
feature
autoclavable
manufacturer/tradename
BRAND
General description
Hollow, with sealing ribs and square grip.
Other Notes
BR144430 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)