Hlavní dokumenty
Z255998
Gum rubber laboratory tubing
tubing I.D. × O.D. 1/4 in. × 1 in.
Synonyma:
vacuum tubing
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
description
heavy wall
packaging
pkg of 10 feet
tubing I.D. × O.D.
1/4 in. × 1 in.
wall size
3/8 in.
General description
Tubing is red, and can be used for pressure or vacuum work.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)