Hlavní dokumenty
Z329010
BRAND® magnetic stirring bar, PTFE, cylindrical with pivot ring
L 40 mm, bar diam. 8 mm
About This Item
UNSPSC Code:
41103806
eCl@ss:
32100100
NACRES:
NB.72
material
cylindrical PTFE-coated
packaging
pack of 10
manufacturer/tradename
BRAND 137430
L
40 mm
bar diam.
8 mm
ring diam.
8.5 mm
General description
For vessels with slightly uneven bottoms. The pivot ring reduces the contact surface and enables the stirrer to adopt the optimum stirring position.
Other Notes
BR137430 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)