Hlavní dokumenty
Z322695
BRAND® beaker, PTFE, low form
capacity 250 mL
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
material
PTFE
feature
graduations: no
manufacturer/tradename
BRAND 90348
H × diam.
95 mm × 70 mm
capacity
250 mL
General description
Excellent thermal and chemical resistance. With reinforced rim and spout, without graduations.
Other Notes
Browse all plastic beakers
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)