Key Documents
GE18-1130-05
EPS power supply, 3501 XL
H × W × D 95 mm × 250 mm × 315 mm, 1 min to 500 h continuous, AC/DC input 102 - 120 V AC
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
feature
wetted part: no
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 18113005
parameter
0-0.95 humidity limit
1.06 bar (according to GPR, maximum alltitude of 2000 m)
4 °C min. temp.
40 °C max. temp.
H × W × D
95 mm × 250 mm × 315 mm
output
1-200 W
Související kategorie
General description
EPS Power Supplies cover the wide range of electrophoresis and blotting applications. Designed for performance, EPS Power Supplies can be relied on for security and reproducibility
All EPS Power Supplies offer Easy-to-read digital display. Timed or continuous runs, with end-of-run alarm, if desired. Automatic parameter limit crossover to prevent overheating and protect experiments and equipment. Automatic recovery after power failure. Membrane keypads for easy programming. Two pairs of power outlets for duplicate parallel runs.
General guidelines for using power supplies: The constant current mode is recommended for blotting to prevent overheating. The constant voltage mode is recommended for other applications, such as nucleic acid submarine and mini-vertical gel electrophoresis. When connecting two electrophoresis units to the same power supply, twice as much current is required at the same voltage. To maintain a constant temperature without using a thermostatic circulator, use the constant power mode.
Storage Class