Hlavní dokumenty
EP5895171001
Rotor for Eppendorf® 5910 R Centrifuge
S-4x500, without buckets
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 4 x 500 mL rectangular buckets
to hold 4 x MTP/flex buckets
S-4x500, without buckets
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5895171001
parameter
4,000 max. RCF (3,220 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5910R
General description
Rotor for Eppendorf(R)5910R Centrifuge
Legal Information
Eppendorf is a registered trademark of Eppendorf AG