Hlavní dokumenty
EP5810751004
Eppendorf® Adapter for 250 mL Rectangular Bucket
holds 16 x 1.5-2 mL tubes, set of 2 pieces
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
holds 16 x 1.5-2 mL tubes
packaging
set of 2 pieces
manufacturer/tradename
Eppendorf® 5810751004
technique(s)
centrifugation: suitable
compatibility
for use with 250 mL reactangular bucket
General description
Tube adapter for Eppendorf 250 mL rectangular bucket.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)