Hlavní dokumenty
EP5810721008
Eppendorf® Adapter for 500 mL Rectangular Bucket
holds 16x7-17 mL tubes, pack of 2 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.78
feature
holds 16x7-17 mL tubes
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5810721008
technique(s)
centrifugation: suitable
compatibility
for use with 500 mL rectangular buckets
General description
Adapter for use with Eppendorf 500 mL rectangular bucket.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)