Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
4 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 3 880,00 Kč |
O této položce
3 880,00 Kč
material
flat bottom bottle, polypropylene copolymer (with sealing closure), translucent bottle
joint
Joints threaded neck (58)
feature
RCF 27,500 × g, with cap
packaging
pack of 4 ea
manufacturer/tradename
Nalgene® 3141-0250
parameter
27500 × g max. RCF
nominal capacity
250 mL
General description
Features and Benefits
- Sturdy polypropylene copolymer (PPCO).
- Designed for low- to moderate-centrifugation speed, large-volume, biological and chemical samples.
- Ideal for use in cell culture harvesting and protein biochemistry applications, including ammonium sulfate precipitations.
- Features a leakproof sealing closure for sample protection.
- Autoclave safe.
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description nominal capacity 250 mL | description nominal capacity 500 mL | description nominal capacity 500 mL | description nominal capacity 250 mL |
| joint Joints threaded neck (58) | joint Joints threaded neck (52.6) | joint Joints threaded neck (63) | joint Joints threaded neck (46.5) |
| manufacturer/tradename Nalgene® 3141-0250 | manufacturer/tradename Nalgene® 3140-0500 | manufacturer/tradename Nalgene® 3141-0500 | manufacturer/tradename Nalgene® 3140-0250 |
| feature with cap, RCF 27,500 × g | feature RCF 13,700 × g, with cap | feature with cap, RCF 13,700 × g | feature with cap, RCF 27,500 × g |
| material polypropylene copolymer (with sealing closure), flat bottom bottle, translucent bottle | material transparent polycarbonate (with sealing closure), polypropylene bottle, screw top cap, round bottle, transparent bottle | material polypropylene copolymer (with sealing closure), round bottle, translucent bottle, flat bottom bottle | material screw top cap, round bottle, transparent bottle, polypropylene bottle, transparent polycarbonate (with sealing closure) |
| parameter 27500 × g max. RCF | parameter 13700 × g max. RCF | parameter 13700 × g max. RCF | parameter 27500 × g max. RCF |
| packaging pack of 4 ea | packaging pack of 4, case of 24 | packaging pack of 4 ea | packaging pack of 4 |
Still not finding the right product?
Explore all of our products under Nalgene® centrifuge bottles, wide-mouth with sealing caps Style 3141
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?