Key Documents
EP5702746002
Eppendorf® Fixed-angle rotor
F-45-24-11, without lid
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 24 x 1.5/2.0 mL tubes
F-45-24-11, without lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5702746002
parameter
4,400 max. RCF (1,770 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5702/5702R/5702RH
Legal Information
Eppendorf is a registered trademark of Eppendorf AG