Key Documents
EP5416301001
Rotor Key For Eppendorf® Rotors
standard
Synonyma:
Rotor Key For Eppendorf® Rotors
About This Item
UNSPSC Code:
41103900
NACRES:
NB.82
feature
standard
manufacturer/tradename
Eppendorf® 5416301001 (Europe)
General description
Centrifuge rotor key for Eppendorf models 5418, 5424 and 5430.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG