Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 12 100,00 Kč |
O této položce
12 100,00 Kč
sterility
non-sterile
packaging
case of 1, pack of 1
manufacturer/tradename
Corning 4515-8L
technique(s)
cell culture: suitable
General description
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description paddle assembly for 8L Proculture spinner flask w/ direct drive overhead motor, 1/cs | description paddle assembly for 36L Proculture spinner flask w/ direct drive overhead motor, 1/cs | description paddle assembly for 15L Proculture spinner flask w/ direct drive overhead motor, 1/cs | description direct overhead drive, 100 mm center cap assembly, 1/cs |
| packaging case of 1, pack of 1 | packaging case of 1, pack of 1 | packaging case of 1, pack of 1 | packaging pack of 1, case of 2 |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| manufacturer/tradename Corning 4515-8L | manufacturer/tradename Corning 4515-36L | manufacturer/tradename Corning 4515-15L | manufacturer/tradename Corning 402614 |
Still not finding the right product?
Explore all of our products under Corning® direct overhead drive assembly for ProCulture® spinner flasks
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?