O této položce
NACRES:
NB.43
UNSPSC Code:
41121804
eCl@ss:
32040501
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlisterility
non-sterile
packaging
pack of 1, case of 2
manufacturer/tradename
Corning 402614
technique(s)
cell culture: suitable
General description
100mm center cap assembly for Corning® Proculture® spinner flasks is designed for use with direct overhead drive motor. These cap assemblies required optional 8L, 15L or 36L paddle assemblies and can be used on all 4500 and 4510 series vertical sidearm flasks.
Legal Information
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries
Still not finding the right product?
Explore all of our products under Corning® direct overhead drive assembly for ProCulture® spinner flasks
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?