Hlavní dokumenty
CLS285522
Corning® cover glasses
square, No. 2, W × L 22 mm × 22 mm
Synonyma:
coverslips, glass covers, slide covers
About This Item
UNSPSC Code:
41121800
NACRES:
NB.16
material
borosilicate glass
square
packaging
pack of 100 ea
case of 1000 ea
manufacturer/tradename
Corning 2855-22
W × L
22 mm × 22 mm
binding type
non-treated surface
Features and Benefits
  • Corning® cover glasses are made from optically clear borosilicate glass.
  • Chemically resistant cover glasses are not affected by weathering or surface attachment.
Legal Information
Corning is a registered trademark of Corning, Inc.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.
Zákazníci si také prohlíželi
Slide 1 of 1
1 of 1
BRAND® cover glass L × W 20 mm × 20 mm, square
BR470050
BRAND® cover glass