Key Documents
ZSR240PFT0
Milli-Q
Milli-Q® SP 240 Prefilter Module
greener alternative
Pretreatment of potable tap water to feed the pure RO water production station
About This Item
UNSPSC Code:
41104200
eCl@ss:
33050190
NACRES:
NC.75
AC/DC input
100 V / 240 V, 50-60 Hz
packaging
pkg of 1 unit
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
parameter
6 kg operating weight (13.2 lb)
system H × W × depth
50.3 cm (19.8 in.) × 14.1 cm (5.6 in.) × 22 cm (8.7 in.)
greener alternative category
General description
To learn how Milli-Q® SQ 2Series water systems work and read a full overview, visit our dedicated page.
The Milli-Q® SP 240 Prefilter Module pretreats potable tap water to feed the pure RO water production station. This prefilter module is recommended to boost the performance of the RO water production station of the Milli-Q® SQ 2Series of water purification systems.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.
Features and Benefits
  • Pump regulates feed water pressure to ensure the RO membrane constant up to 42 L/h high flow rate
  • SQPAK Gard pretreatment pack protects the RO membrane by efficiently removing chlorine, colloids and particulate contaminants from feed tap water
  • Compact setup and self-guided installation manual and video let you plug the module next to the pure RO water production station, and move it later if needed
  • Individual Twist & Lock cartridge replacement, for each exhausted purification step, simplifies maintenance and limits running cost
  • Compact size supports lab sustainability efforts
Components
The indicated price currently corresponds to the above catalog number only; it is subject to change at any time without notice.
This catalog number does not include:
  • SQPAK cartridge
  • Power cord adapted to the local electrical network
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
SQPAK is a trademark of Merck KGaA, Darmstadt, Germany
accessory
Č. produktu
Popis
Stanovení ceny
Navštívit knihovnu dokumentů
Protokoly
View instructional videos and download User Manuals for Milli-Q® SQ 2Series lab water systems. You’ll be up and running in no time. Buy online systems, accessories, cartridges and services.
Prohlédněte si instruktážní videa a stáhněte si uživatelské příručky k laboratorním vodním systémům Milli-Q® SQ 2Series. Během chvilky budete moci začít pracovat. Nakupujte online systémy, příslušenství, kazety a služby.
Související obsah
Discover a new way to think about ultrapure water. Now any lab bench with a plug can freshly dispense 18.2 MΩ∙cm ultrapure water. Robust Quality. Simple. Compact. Scalable. A Greener Alternative Product. Buy Online.
Objevte nový způsob uvažování o ultračisté vodě. Nyní může každý laboratorní stůl se zástrčkou čerstvě dávkovat ultračistou vodu o koncentraci 18,2 MΩ∙cm. Robustní kvalita. Jednoduché. Kompaktní. Škálovatelná. Ekologičtější alternativní produkt. Koupit online.