Hlavní dokumenty
NA12/21
Millipore
NovAseptic Valve, Diaphragm, Silicone
About This Item
UNSPSC Code:
40141600
eCl@ss:
37051001
material
valve (Diaphragm, Silicone)
description
Smooth (hydrophobic)
parameter
-1 to -6 bar pressure
weight
0.01 kg0.022 lb
Application
The applied valve body and actuator may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Packaging
The Diaphragm is packaged in a closed box.
Other Notes
Short term use: min -50 °C; max Dry Heat (oxygen rich environment) 220 °C; max Dry Heat (oxygen poor environment) 150 °C; max Steam 130 °C.
Disclaimer
All Diaphragms are manufactured with materials in compliance with FDA regulations §177.2600 and supplied with statement. The Diaphragm meets USP Biological Tests for Plastics, Class VI.