Hlavní dokumenty
GFDX083000
Millipore
Glass Fiber Diagnostic Pad
filter W × L 8 mm × 300 mm, pack of 100 strips
Synonyma:
Conjugate pad, Diagnostic pad, Glass Fiber Diagnostic Pad 8 mm x 300 mm strips 100pk, Glass fiber pad
About This Item
UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
packaging
pack of 100 strips
Quality Level
manufacturer/tradename
SureWick®
W
8 mm
filter W × L
8 mm × 300 mm
shipped in
ambient
General description
SureWick® Pad Materials are designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance. SureWick® glass fiber conjugate / diagnostic pads are used to release detector reagents and faciltiate sample flow across the lateral flow membrane. This pack contains 100 strips of 8mm x 300mm glass fiber diagnostic pad (GFDX) material.
Application
SureWick® Pad Materials are optimised for use as sample, conjugate, or absorbent pads in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs etc.), as well as in women’s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing, and animal health.
Features and Benefits
  • SureWick® glass fiber pads have low extractables and excellent consistency.
  • They provide reliable, predictable sample flow and reagent delivery.
  • They exhibit consistent thickness, bed volume, and uniform surface quality.
  • Quick Delivery and Lead time.
  • Excellent Technical support.
Legal Information
SUREWICK is a registered trademark of Merck KGaA, Darmstadt, Germany
pictograms
Health hazard
signalword
Danger
hcodes
Hazard Classifications
Carc. 1B Inhalation
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c