Key Documents
Z741033
Sigma-Aldrich
CJ Tee Syringe Adapter - CGA 180 Inlet
About This Item
UNSPSC Code:
12142200
NACRES:
NA.12
Quality Level
Application
Use to extract samples from gas cylinders. Please see attached bulletin for detailed instructions.