Key Documents
Z740682
Sigma-Aldrich
Aldrich® Essentials filter flask, with side arm
capacity 500 mL
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass flask
flat bottom flask
joint
joint stopper7
packaging
pkg of 12 ea
capacity
500 mL
General description
Clear borosilicate glass Erlenmeyer style flask with side arm for connecting to vacuum or aspirator line. Used for filtration with Buchner funnels, Gooch crucibles, etc. Beaded rim for rubber gromets or filter adapters or rubber stoppers.