Key Documents
Z646253
Sigma-Aldrich
Aldrich® drum fittings
vented faucet for 5/10 L Aldrich drums
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
diam.
50 mm
General description
Robust PP drum fittings designed to fit Aldrich 5, 10, 20, 30 & 60 L plastic drums, making chemical dispensing from our containers safe and simple.