Hlavní dokumenty
Z175633
Sigma-Aldrich
Silicone insulating tape
roll W 1 in. × 12 yard, red
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
sterility
non-sterile
manufacturer/tradename
Sigma-Aldrich
roll W
1 in. × 12 yard
color
red
General description
Shape-conforming, fuses to itself. Insulates against moisture, heat, and cold. No adhesive residue left upon removal. Useful temperature range: -65 to 260°C.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)