Hlavní dokumenty
Z645222
IKA® M 22 hard metal cutter
M 22 hard metal cutter, tungsten carbide, 1/cs
About This Item
UNSPSC Code:
41121800
NACRES:
NB.74
material
tungsten carbide
packaging
case of 1
pack of 1
manufacturer/tradename
IKA 521800
suitability
suitable for (hard materials up to Mohs hardness 9)
General description
Note: Made of tungsten carbide for hard materials up to Mohs hardness 9.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)