Key Documents
Z562548
LabJaws hook lattice rod connector
jaw capacity 0-13 mm , 0-51 inches, nickel plated (zinc)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
material
nickel plated (zinc)
manufacturer/tradename
Troemner 916128
jaw capacity
0-13 mm , 0-51 inches
General description
Corrosion resistant. The hook allows for one-handed assembly of two components with one adjustment screw.
Legal Information
LabJaws is a trademark of Henry Troemner LLC