Key Documents
Z552496
Sigma-Aldrich
Aldrich® glass thermometer well
fits flask, 50 L, immersion L 350 mm
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
joint ST/NS24/40
feature
fits flask 50 L
Související kategorie
General description
Knobbed on sides for easy handling.