Key Documents
Z552453
Sigma-Aldrich
Aldrich® glass thermometer well
fits flask, 5 L, immersion L 160 mm
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
joint ST/NS24/40
feature
fits flask 5 L
General description
Knobbed on sides for easy handling.