Z547786
SafetyBarb® hose connector
straight
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Doporučené produkty
General description
Consists of PBT cap, PP barbed connector 8.5 mm O.D., and silicone rubber seal. GL14 thread.
Legal Information
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)