This adapter is a 1/4 inch hose barb fitting to a male Luer Slip fitting, which is 4mm (approximately 1/8 inch) at the opening, with a 6 % taper. Please note that this fitting is compatible only with a female Luer Slip fitting. The Luer fitting has no barbs and is therefore not suitable for the connection of flexible tubing.
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
10 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 2 110,00 Kč |
O této položce
UNSPSC Code:
41104008
NACRES:
NB.21
2 110,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlipackaging
pkg of 10 ea
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
General description
Male Luer Fitting for 1/4 in. Tubing is an accessory for use with TDS3 Thermal Desorption Storage Containers.
1 of 1
Tato položka | |||
|---|---|---|---|
| description pkg of 10 ea | description pkg of 20 ea | description pkg of 20 ea | description for use with detector tube |
| technique(s) active air sampling: suitable | technique(s) active air sampling: suitable | technique(s) active air sampling: suitable | technique(s) active air sampling: suitable |
| packaging pkg of 10 ea | packaging pkg of 20 ea | packaging pkg of 20 ea | packaging - |
| application(s) air monitoring | application(s) air monitoring | application(s) air monitoring | application(s) air monitoring |
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?
-
What is the size (in inches) of the other side of this adapter? It is designed for 1/4 inch tubing, however I am wondering about the outer diameter of the opposite end. Thank you!