Hlavní dokumenty
Z516236
Sigma-Aldrich
Aldrich® side-arm distilling head
joint: ST/NS 29/32, thermometer joint: ST/NS 10/30
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
joint
thermometer joint: ST/NS10/30
joint: ST/NS29/32
side arm L
225 mm
General description
A 50mm immersion length thermometer is recommended for the 14/20 joint, and 75mm immersion length thermometers are recommended for the 24/40 and 29/32 joints.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)