Key Documents
Z507121
Supelco
Michel-Miller column plug
Front seal, PTFE, thread size 80 mm
About This Item
UNSPSC Code:
12000000
NACRES:
SB.52
material
PTFE
manufacturer/tradename
Ace Glass 584660
technique(s)
LPLC: suitable
thread size
80 mm
General description
Solid plug for use as stopper with Ace-Threds. Seals M-M column ends, Plugs with #50 Ace-Thred fit Ace large columns.