Z251860
Michel-Miller column stopper
Ace-Thred for #15
manufacturer/tradename
Ace Glass 580311
technique(s)
LPLC: suitable
fitting
Ace-Thred for #15
General description
PTFE for use at either end of any chromatography column with Ace-Threds. Makes a leak-tight seal without O-rings.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)