Key Documents
Z405663
J. Young needle valve
PTFE, side arm O.D. 7 mm
About This Item
UNSPSC Code:
40141602
NACRES:
NB.79
material
PTFE
manufacturer/tradename
J. Young
side arm O.D.
7 mm
General description
• PTFE piston with unique PTFE-sheathed elastomer O-ring seals
• PTFE needle shut-off point
• Calibrated barrel indicates flow and facilitates reproducibility