Z403083
Büchi® receiving flask
capacity 2,000 mL, plastic-coated glass
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Doporučené produkty
material
plastic-coated glass
round bottom
single-neck
joint
joint SJ35/20
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Büchi 25265
capacity
2,000 mL
Související kategorie
General description
These fit all Rotavapor models.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)