Key Documents
Z402974
Sigma-Aldrich
Büchi® evaporating flask, pear-shaped
volume 250 mL, glass, joint: ST/NS 24/40
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
glass
pear-shaped
single-neck
joint
joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Büchi 08754
capacity
250 mL
volume
250 mL
General description
These flasks fit R200/205, R210/215, and RII evaporators
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG