Key Documents
Z281395
Sigma-Aldrich
Sampler brush kit
pkg of 1 ea
About This Item
UNSPSC Code:
47131605
NACRES:
NB.35
packaging
pkg of 1 ea
General description
Convenience pack samples the following high quality laboratory brushes: 1 Micro tube, 1 Flask, 1 Test tube, 1 Burette, 1 Cylinder, 1 Pipette, 1 Narrow tube, 1 Funnel.
Zákazníci si také prohlíželi
Slide 1 of 5
1 of 5
Laboratory brushes Funnel, L 13 in., brush diam. × L 2 1/4 in. × 6 in.
Z281336
Laboratory brushes
Laboratory brushes Cylinder, L 11 in., brush diam. × L 2 3/8 in. × 5 in.
Z281271
Laboratory brushes
Laboratory brushes Centrifuge, L 12 in., brush diam. × L 1 1/2 in. × 4 1/2 in.
Z281352
Laboratory brushes
Laboratory brushes Test tube, L 10 in., brush diam. × L 1 3/8 in. × 4 in.
Z281263
Laboratory brushes
Laboratory brushes Burette, L 36 in., brush diam. × L 3/4 in. × 3 1/4 in.
Z281379
Laboratory brushes