Key Documents
Z281271
Laboratory brushes
Cylinder, L 11 in., brush diam. × L 2 3/8 in. × 5 in.
About This Item
UNSPSC Code:
47131605
NACRES:
NB.35
agency
suitable for EPA OTM-45
packaging
pkg of 4 ea
L
11 in.
brush diam. × L
2 3/8 in. × 5 in.
General description
Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water. FDA and USDA approved materials.
Zákazníci si také prohlíželi
Související obsah
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.