Z247529
Coors™ porcelain pestle
for Z247480
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
Doporučené produkty
material
porcelain
packaging
pack of 1 ea
manufacturer/tradename
Coorstek 66320
pestle L
180 mm
Související kategorie
General description
Oversized pestle glazed to grinding surface. Order mortar separately.
Application
The Coors™ porcelain pestle is suitable to use for crushing, grinding, and mixing.
Legal Information
Coors is a trademark of Coorstek, Inc.