Key Documents
Z238759
PTFE blade for stirrer shafts
blade diam. 125 mm
About This Item
UNSPSC Code:
41103800
NACRES:
NB.72
material
PTFE
packaging
pack of 1 ea
manufacturer/tradename
Cowie 002.1125.1
blade diam.
125 mm