Key Documents
Z238732
PTFE blade for stirrer shafts
blade diam. 75 mm
About This Item
UNSPSC Code:
41103800
NACRES:
NB.72
material
PTFE
packaging
pack of 1 ea
manufacturer/tradename
Cowie 002.1075.1
blade diam.
75 mm
Zákazníci si také prohlíželi
Slide 1 of 1
1 of 1
PTFE stirrer shaft shaft diam. 9.5 mm, L 650 mm
Z238716
PTFE stirrer shaft